missing translation for 'onlineSavingsMsg'
Learn More

Digoxin, 95%

Catalog No. AC228120050
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC228120050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC228120050 Supplier Thermo Scientific Chemicals Supplier No. 228120050

CAS: 20830-75-5 | C41H64O14 | 780.94 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 20830-75-5
Molecular Formula C41H64O14
Molecular Weight (g/mol) 780.94
MDL Number MFCD00003674
InChI Key LTMHDMANZUZIPE-PUGKRICDSA-N
Synonym digoxin, 12beta-hydroxydigitoxin, digoxine, lanoxin, lanoxicaps, digosin, digossina, digoxina, digoxinum, lanoxicaps tn
PubChem CID 2724385
ChEBI CHEBI:4551
IUPAC Name 3-[(3S,5R,8R,9S,10S,12R,13S,14S,17R)-3-[(2R,4S,5S,6R)-5-[(2S,4S,5S,6R)-5-[(2S,4S,5S,6R)-4,5-dihydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyloxan-2-yl]oxy-12,14-dihydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,15,16,17-tet
SMILES CC1C(C(CC(O1)OC2C(OC(CC2O)OC3C(OC(CC3O)OC4CCC5(C(C4)CCC6C5CC(C7(C6(CCC7C8=CC(=O)OC8)O)C)O)C)C)C)O)O

Specifications

Melting Point 248.0°C to 250.0°C
Assay Percent Range 94% min. (on dried substance) (HPLC)
Packaging Glass bottle
Quantity 5 g
Merck Index 15,3186
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in diluted alcohol,pyridine,mixture of,chloroform and alcohol,almost insoluble in ether,acetone,ethylacetate,,chloroform
Formula Weight 780.94
Percent Purity 95%
Chemical Name or Material Digoxin
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Fatal if swallowed.
Precautionary Statement GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
R
EINECSNumber 244-068-1

RUO – Research Use Only