missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diglycolic Acid 98.0+%, TCI America™
Supplier: TCI America D0545500G
Specifications
| Diglycolic Acid | |
| 110-99-6 | |
| C4H6O5 | |
| diglycolic acid, 2,2'-oxydiacetic acid, oxydiacetic acid, acetic acid, 2,2'-oxybis, oxybisacetic acid, oxydiethanolic acid, diglycolicacid, bis carboxymethyl ether, oxodiacetic acid, 3-oxapentanedioic acid | |
| OC(=O)COCC(O)=O | |
| 134.09 | |
| 134.09 | |
| 500 g |
| 144°C | |
| White | |
| MFCD00004309 | |
| QEVGZEDELICMKH-UHFFFAOYSA-N | |
| 2-(carboxymethoxy)acetic acid | |
| 8088 | |
| ≥98.0% (GC,T) | |
| Crystalline Powder |
Chemical Identifiers
| 110-99-6 | |
| 134.09 | |
| QEVGZEDELICMKH-UHFFFAOYSA-N | |
| 8088 | |
| OC(=O)COCC(O)=O |
| C4H6O5 | |
| MFCD00004309 | |
| diglycolic acid, 2,2'-oxydiacetic acid, oxydiacetic acid, acetic acid, 2,2'-oxybis, oxybisacetic acid, oxydiethanolic acid, diglycolicacid, bis carboxymethyl ether, oxodiacetic acid, 3-oxapentanedioic acid | |
| 2-(carboxymethoxy)acetic acid |
Safety and Handling
EINECSNumber : (2)-2748&(2)-3080&(2)-3435
RTECSNumber : AJ1530000
TSCA : Yes