missing translation for 'onlineSavingsMsg'
Learn More

Diglycolic Acid 98.0+%, TCI America™

Catalog No. p-7126277
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D054525G 25 g
D0545500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D054525G Supplier TCI America Supplier No. D054525G
Only null left

Chemical Identifiers

CAS 110-99-6
Molecular Formula C4H6O5
Molecular Weight (g/mol) 134.09
MDL Number MFCD00004309
InChI Key QEVGZEDELICMKH-UHFFFAOYSA-N
Synonym diglycolic acid, 2,2'-oxydiacetic acid, oxydiacetic acid, acetic acid, 2,2'-oxybis, oxybisacetic acid, oxydiethanolic acid, diglycolicacid, bis carboxymethyl ether, oxodiacetic acid, 3-oxapentanedioic acid
PubChem CID 8088
IUPAC Name 2-(carboxymethoxy)acetic acid
SMILES OC(=O)COCC(O)=O

Specifications

Melting Point 144°C
Color White
Formula Weight 134.09
Percent Purity ≥98.0% (GC,T)
Quantity 25 g
Physical Form Crystalline Powder
Chemical Name or Material Diglycolic Acid
EINECSNumber (2)-2748&(2)-3080&(2)-3435
RTECSNumber AJ1530000
TSCA Yes