missing translation for 'onlineSavingsMsg'
Learn More

Diglycidyl 1,2-Cyclohexanedicarboxylate (so called), TCI America™

Catalog No. C1434500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C1434500G 500 g
C143425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C1434500G Supplier TCI America Supplier No. C1434500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 5493-45-8
Molecular Formula C14H20O6
Molecular Weight (g/mol) 284.308
MDL Number MFCD00128914
InChI Key XFUOBHWPTSIEOV-UHFFFAOYSA-N
Synonym 1,2-Cyclohexanedicarboxylic Acid Diglycidyl Ester
PubChem CID 21660
IUPAC Name bis(oxiran-2-ylmethyl) cyclohexane-1,2-dicarboxylate
SMILES C1CCC(C(C1)C(=O)OCC2CO2)C(=O)OCC3CO3

Specifications

Color Yellow
Boiling Point 205°C
Quantity 500 g
Formula Weight 284.31
Physical Form Liquid
Chemical Name or Material Diglycidyl 1,2-Cyclohexanedicarboxylate (so called)
RTECSNumber GU9070000
TSCA Yes