accessibility menu, dialog, popup

UserName

CAS RN 5493-45-8

IUPAC Name: 1,2-bis[(oxiran-2-yl)methyl] cyclohexane-1,2-dicarboxylate
Synonyms: 1,2-bis(oxiran-2-ylmethyl) cyclohexane-1,2-dicarboxylate
Molecular Weight (g/mol): 284.31
Molecular Formula: C14H20O6
InChi Key: XFUOBHWPTSIEOV-UHFFFAOYNA-N
SMILES: O=C(OCC1CO1)C1CCCCC1C(=O)OCC1CO1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results