missing translation for 'onlineSavingsMsg'
Learn More

Diglycerol 80.0+%, TCI America™

Catalog No. T011925G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T011925G 25 g
T0119500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T011925G Supplier TCI America Supplier No. T011925G
Only null left

Chemical Identifiers

CAS 627-82-7
Molecular Formula C6H14O5
Molecular Weight (g/mol) 166.173
MDL Number MFCD00049316
InChI Key GPLRAVKSCUXZTP-UHFFFAOYSA-N
Synonym diglycerol, diglycerine, diglycerin, alpha,alpha'-diglycerol, glyceryl ether, 1,2-propanediol, 3,3'-oxybis, oxydi propanediol, tetrahydroxydipropyl ether, .alpha.,.alpha.'-diglycerol, 3-2,3-dihydroxypropoxy propane-1,2-diol
PubChem CID 42953
IUPAC Name 3-(2,3-dihydroxypropoxy)propane-1,2-diol
SMILES C(C(COCC(CO)O)O)O

Specifications

Color Yellow
Quantity 25 g
Formula Weight 166.17
Percent Purity ≥80.0% (GC)
Physical Form Liquid
Chemical Name or Material Diglycerol
EINECSNumber (2)-0418
TSCA No