accessibility menu, dialog, popup

UserName

CAS RN 627-82-7

HOOHOOHOH

CAS RN 627-82-7

IUPAC Name: 3-(2,3-dihydroxypropoxy)propane-1,2-diol
Synonyms: 3-(2,3-dihydroxypropoxy)propane-1,2-diol
Molecular Weight (g/mol): 166.17
Molecular Formula: C6H14O5
InChi Key: GPLRAVKSCUXZTP-UHFFFAOYNA-N
SMILES: OCC(O)COCC(O)CO
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results