missing translation for 'onlineSavingsMsg'
Learn More

Digitoxigenin 97.0+%, TCI America™

Catalog No. D054110MG
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D054110MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D054110MG Supplier TCI America Supplier No. D054110MG

Chemical Identifiers

CAS 143-62-4
Molecular Formula C23H34O4
Molecular Weight (g/mol) 374.521
MDL Number MFCD00003687
InChI Key XZTUSOXSLKTKJQ-CESUGQOBSA-N
PubChem CID 4369270
ChEBI CHEBI:42219
IUPAC Name 3-[(3S,5R,8R,9S,10S,13R,14S,17R)-3,14-dihydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,15,16,17-tetradecahydrocyclopenta[a]phenanthren-17-yl]-2H-furan-5-one
SMILES CC12CCC(CC1CCC3C2CCC4(C3(CCC4C5=CC(=O)OC5)O)C)O

Specifications

Melting Point 253°C
Color White
Quantity 10 mg
UN Number 2811
Formula Weight 374.52
Percent Purity ≥97.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Digitoxigenin
RTECSNumber FH4975000
TSCA No