accessibility menu, dialog, popup

UserName

H3C(S)(S)OH(R)(R)(S)(R)H3C(R)(S)OHOO

CAS RN 143-62-4

IUPAC Name: 4-[(1R,3aS,3bR,5aR,7S,9aS,9bS,11aR)-3a,7-dihydroxy-9a,11a-dimethyl-hexadecahydro-1H-cyclopenta[a]phenanthren-1-yl]-2,5-dihydrofuran-2-one
Synonyms: digitoxigenin digitoxigenine echujetin uzarigenin thevetigenin cerberigenin evonogenin
Molecular Weight (g/mol): 374.52
Molecular Formula: C23H34O4
InChi Key: XZTUSOXSLKTKJQ-CESUGQOBSA-N
SMILES: C[C@]12CC[C@H]3[C@@H](CC[C@@H]4C[C@@H](O)CC[C@]34C)[C@@]1(O)CC[C@@H]2C1=CC(=O)OC1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results