missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Digestion Reagent, with Mercury Catalyst, Ricca Chemical
For Analysis of Nitrogen (Kjeldahl)
Supplier: Ricca Chemical Company RI255016
Alert:
The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.
Chemical Identifiers
| 65.0, 24.42, 8.45, 0.13 | |
| 68.0, 25.42, 8.8, 0.14 | |
| 7664-93-9, 7778-80-5, 21908-53-2 | |
| Digestion Reagent, With Mercury Catalyst |
Specifications
| Digestion Reagent | |
| Colorless | |
| 65.0, 24.42, 8.45, 0.13 | |
| 68.0, 25.42, 8.8, 0.14 |
| 500 mL | |
| With Mercury Catalyst | |
| Natural Poly Bottle | |
| 7664-93-9 , 7778-80-5 , 21908-53-2 |