Learn More
Digestion Reagent, with Mercury Catalyst, Ricca Chemical
For Analysis of Nitrogen (Kjeldahl)
Supplier:  Ricca Chemical Company 25501
 View more versions of this product
Catalog No. 25501
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Alert:
The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.
Description
Description
Specifications
Chemical Identifiers
65.0, 24.42, 8.45, 0.13
68.0, 25.42, 8.8, 0.14
7664-93-9, 7778-80-5, 21908-53-2
Digestion Reagent, With Mercury Catalyst
Specifications
Digestion Reagent
Colorless
65.0, 24.42, 8.45, 0.13
68.0, 25.42, 8.8, 0.14
4 L
With Mercury Catalyst
Natural Poly Bottle
7664-93-9 , 7778-80-5 , 21908-53-2
Product Suggestions
Product Suggestions