missing translation for 'onlineSavingsMsg'
Learn More

Digestion Reagent, with Copper Catalyst, Ricca Chemical

Catalog No. 25511
Change view
Click to view available options
Quantity:
4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
25511 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 25511 Supplier Ricca Chemical Company Supplier No. RI25511

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For Analysis of Nitrogen (Kjeldahl)

Specifications

Chemical Name or Material Digestion Reagent
Name Note With Copper Catalyst
CAS Min % 0.51, 23.95
Color Blue
CAS Max % 8.81, 0.53
CAS 7778-80-5, 7664-93-9, 7732-18-5, 7758-99-8
Physical Form Liquid
Quantity 4 L
Packaging Natural Poly Bottle
Grade Laboratory