missing translation for 'onlineSavingsMsg'
Learn More

Difurfuryl Disulfide 95.0+%, TCI America™

Catalog No. D138425G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D138425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D138425G Supplier TCI America Supplier No. D138425G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 4437-20-1
Molecular Formula C10H10O2S2
Molecular Weight (g/mol) 226.31
MDL Number MFCD00010082
InChI Key CBJPZHSWLMJQRI-UHFFFAOYSA-N
Synonym Furfuryl Disulfide
PubChem CID 20499
IUPAC Name 2-({[(furan-2-yl)methyl]disulfanyl}methyl)furan
SMILES C(SSCC1=CC=CO1)C1=CC=CO1

Specifications

Color Yellow
Boiling Point 121°C
Quantity 25 g
Formula Weight 226.31
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Difurfuryl Disulfide
EINECSNumber (5)-0052
TSCA Yes
Recommended Storage Refrigerator