accessibility menu, dialog, popup

UserName

SSOO

CAS RN 4437-20-1

IUPAC Name: 2-({[(furan-2-yl)methyl]disulfanyl}methyl)furan
Synonyms: 2-{[(furan-2-ylmethyl)disulfanyl]methyl}furan
Molecular Weight (g/mol): 226.31
Molecular Formula: C10H10O2S2
InChi Key: CBJPZHSWLMJQRI-UHFFFAOYSA-N
SMILES: C(SSCC1=CC=CO1)C1=CC=CO1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results