missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DiFMU (6,8-Difluoro-7-Hydroxy-4-Methylcoumarin), reference standard
Description
DIFMU can be used as a reference standard for ether, ester and phosphate substrates derived from this blue-fluorescent dye.
Specifications
Specifications
| Quantity | 10 mg |
| Product Type | Reference Standard |
| Content And Storage | Store at room temperature and protect from light. |
| Shipping Condition | Room Temperature |
| Label or Dye | DiFMU |
| Label Type | Coumarin Dyes |
For Research Use Only. Not for use in diagnostic procedures.