missing translation for 'onlineSavingsMsg'
Learn More

Difluoromethyl Phenyl Sulfone 98.0+%, TCI America™

Catalog No. p-7128449
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D47641G 1 g
D47645G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D47641G Supplier TCI America Supplier No. D47641G
Only null left

Chemical Identifiers

CAS 1535-65-5
Molecular Formula C7H6F2O2S
Molecular Weight (g/mol) 192.18
MDL Number MFCD01050170
InChI Key LRHDNAVPELLXDL-UHFFFAOYSA-N
Synonym difluoromethyl sulfonyl benzene, difluoromethyl phenyl sulfone, difluoromethanesulfonylbenzene, phenyl difluoromethyl sulfone, benzene, difluoromethyl sulfonyl, difluoromethyl sulfonylbenzene, difluoro-methanesulfonyl-benzene, difluoromethyl sulfonyl-benzene
PubChem CID 11816356
IUPAC Name difluoromethylsulfonylbenzene
SMILES C1=CC=C(C=C1)S(=O)(=O)C(F)F

Specifications

Color Colorless
Boiling Point 120°C
Quantity 1 g
Formula Weight 192.18
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Difluoromethyl Phenyl Sulfone