missing translation for 'onlineSavingsMsg'
Learn More

Difluoromethyl 2,2,3,3-Tetrafluoropropyl Ether 98.0+%, TCI America™

Catalog No. p-7128291
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D44721G 1 g
D44725G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D44721G Supplier TCI America Supplier No. D44721G
Only null left

Chemical Identifiers

CAS 35042-99-0
Molecular Formula C4H4F6O
Molecular Weight (g/mol) 182.07
MDL Number MFCD00236119
InChI Key PCTQNZRJAGLDPD-UHFFFAOYSA-N
PubChem CID 2737027
IUPAC Name 3-(difluoromethoxy)-1,1,2,2-tetrafluoropropane
SMILES FC(F)OCC(F)(F)C(F)F

Specifications

Color Colorless
Boiling Point 76°C
Quantity 1 g
UN Number 3271
Formula Weight 182.07
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Difluoromethyl 2,2,3,3-Tetrafluoropropyl Ether