accessibility menu, dialog, popup

UserName

FFFFOFF

CAS RN 35042-99-0

IUPAC Name: 3-(difluoromethoxy)-1,1,2,2-tetrafluoropropane
Synonyms: 3-(difluoromethoxy)-1,1,2,2-tetrafluoropropane
Molecular Weight (g/mol): 182.07
Molecular Formula: C4H4F6O
InChi Key: PCTQNZRJAGLDPD-UHFFFAOYSA-N
SMILES: FC(F)OCC(F)(F)C(F)F
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results