missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Difluoroacetaldehyde Ethyl Hemiacetal 90.0+%, TCI America™
$387.73 - $1165.03
Chemical Identifiers
| CAS | 148992-43-2 |
|---|---|
| Molecular Formula | C4H8F2O2 |
| Molecular Weight (g/mol) | 126.10 |
| MDL Number | MFCD01321160 |
| InChI Key | WEEOMNFWRCDRJI-UHFFFAOYNA-N |
| Synonym | 1-Ethoxy-2,2-difluoroethanol |
| PubChem CID | 2758246 |
| IUPAC Name | 1-ethoxy-2,2-difluoroethan-1-ol |
| SMILES | CCOC(O)C(F)F |
Chemical Identifiers
| 148992-43-2 | |
| 126.10 | |
| WEEOMNFWRCDRJI-UHFFFAOYNA-N | |
| 2758246 | |
| CCOC(O)C(F)F |
| C4H8F2O2 | |
| MFCD01321160 | |
| 1-Ethoxy-2,2-difluoroethanol | |
| 1-ethoxy-2,2-difluoroethan-1-ol |
Specifications
| 148992-43-2 | |
| C4H8F2O2 | |
| 5 g | |
| 1-Ethoxy-2,2-difluoroethanol | |
| CCOC(O)C(F)F | |
| 126.10 | |
| 126.10 | |
| Liquid |
| Colorless | |
| MFCD01321160 | |
| 1993 | |
| WEEOMNFWRCDRJI-UHFFFAOYNA-N | |
| 1-ethoxy-2,2-difluoroethan-1-ol | |
| 2758246 | |
| ≥90.0% (NMR) | |
| Difluoroacetaldehyde Ethyl Hemiacetal |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator