missing translation for 'onlineSavingsMsg'
Learn More

Difluoroacetaldehyde Ethyl Hemiacetal 90.0+%, TCI America™

Catalog No. d252325g
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
5 g
Catalog No. Quantity
D252325G 25 g
D25235G 5 g
2 options

Catalog No. D252325G

Supplier: TCI America D252325G

Only null left

Chemical Identifiers

CAS 148992-43-2
Molecular Formula C4H8F2O2
Molecular Weight (g/mol) 126.10
MDL Number MFCD01321160
InChI Key WEEOMNFWRCDRJI-UHFFFAOYNA-N
Synonym 1-Ethoxy-2,2-difluoroethanol
PubChem CID 2758246
IUPAC Name 1-ethoxy-2,2-difluoroethan-1-ol
SMILES CCOC(O)C(F)F

Specifications

Color Colorless
Quantity 25 g
UN Number 1993
Formula Weight 126.10
Percent Purity ≥90.0% (NMR)
Physical Form Liquid
Chemical Name or Material Difluoroacetaldehyde Ethyl Hemiacetal
TSCA No
Recommended Storage Refrigerator