missing translation for 'onlineSavingsMsg'
Learn More

Diflunisal, Thermo Scientific Chemicals

Catalog No. p-7077860
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC460890050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC460890050 Supplier Thermo Scientific Chemicals Supplier No. 460890050

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 22494-42-4
Molecular Formula C13H8F2O3
Molecular Weight (g/mol) 250.20
MDL Number MFCD00057834
InChI Key HUPFGZXOMWLGNK-UHFFFAOYSA-N
Synonym diflunisal, dolobid, dolobis, fluniget, flovacil, fluodonil, adomal, flustar, 5-2,4-difluorophenyl salicylic acid, diflunisalum
PubChem CID 3059
ChEBI CHEBI:39669
IUPAC Name 5-(2,4-difluorophenyl)-2-hydroxybenzoic acid
SMILES OC(=O)C1=CC(=CC=C1O)C1=CC=C(F)C=C1F

Specifications

Melting Point 207.0°C to 209.0°C
Loss on Drying 0.5% max. (60°C, 4 H) (vacuum)
Infrared Spectrum Authentic
Linear Formula F2C6H3C6H3(OH)COOH
Quantity 5 g
Solubility Information Solubility in water: .. Other solubilities: soluble in methanol, dmso
Formula Weight 250.2
Percent Purity ≥97.5% (HPLC)
Chemical Name or Material Diflunisal
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Suspected of damaging fertility or the unborn child.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep at
EINECSNumber 245-034-9

RUO – Research Use Only