missing translation for 'onlineSavingsMsg'
Learn More

Diflunisal, MilliporeSigma™ Supelco™

Catalog No. 111012181
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111012181 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111012181 Supplier Merck Emd Millipore Supplier No. D32815G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Featured Industries: Pharmaceutical (small molecule)

Specifications

CAS 22494-42-4
Physical Form Neat
Linear Formula C13H8F2O3
MDL Number MFCD00057834
Synonym 2′,4′-Difluoro-4-hydroxy-[1,1′-biphenyl]-3-carboxylic acid; 5-(2,4-Difluorophenyl)salicylic acid
Type Analytical Standard
Quantity 5 g
Formula Weight 250.20
Grade Analytical Standard