missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diflufenican 98.0+%, TCI America™
$249.78 - $741.89
Chemical Identifiers
| CAS | 83164-33-4 |
|---|---|
| Molecular Formula | C19H11F5N2O2 |
| Molecular Weight (g/mol) | 394.301 |
| MDL Number | MFCD01311804 |
| InChI Key | WYEHFWKAOXOVJD-UHFFFAOYSA-N |
| Synonym | diflufenican, n-2,4-difluorophenyl-2-3-trifluoromethyl phenoxy nicotinamide, diflufenicanil, diflufenican bsi:iso, unii-3t8d1vdo6k, n-2,4-difluorophenyl-2-3-trifluoromethyl phenoxy pyridine-3-carboxamide, 3t8d1vdo6k, 2',4'-difluoro-2-alpha,alpha,alpha-trifluoro-m-tolyloxy nicotinanilide, n-2,4-difluorophenyl-2-3-trifluoromethyl phenoxy-3-pyridinecarboxamide |
| PubChem CID | 91735 |
| ChEBI | CHEBI:81824 |
| IUPAC Name | N-(2,4-difluorophenyl)-2-[3-(trifluoromethyl)phenoxy]pyridine-3-carboxamide |
| SMILES | C1=CC(=CC(=C1)OC2=C(C=CC=N2)C(=O)NC3=C(C=C(C=C3)F)F)C(F)(F)F |
Chemical Identifiers
| 83164-33-4 | |
| 394.301 | |
| WYEHFWKAOXOVJD-UHFFFAOYSA-N | |
| 91735 | |
| N-(2,4-difluorophenyl)-2-[3-(trifluoromethyl)phenoxy]pyridine-3-carboxamide |
| C19H11F5N2O2 | |
| MFCD01311804 | |
| diflufenican, n-2,4-difluorophenyl-2-3-trifluoromethyl phenoxy nicotinamide, diflufenicanil, diflufenican bsi:iso, unii-3t8d1vdo6k, n-2,4-difluorophenyl-2-3-trifluoromethyl phenoxy pyridine-3-carboxamide, 3t8d1vdo6k, 2',4'-difluoro-2-alpha,alpha,alpha-trifluoro-m-tolyloxy nicotinanilide, n-2,4-difluorophenyl-2-3-trifluoromethyl phenoxy-3-pyridinecarboxamide | |
| CHEBI:81824 | |
| C1=CC(=CC(=C1)OC2=C(C=CC=N2)C(=O)NC3=C(C=C(C=C3)F)F)C(F)(F)F |
Specifications
| 83164-33-4 | |
| C19H11F5N2O2 | |
| 5 g | |
| WYEHFWKAOXOVJD-UHFFFAOYSA-N | |
| N-(2,4-difluorophenyl)-2-[3-(trifluoromethyl)phenoxy]pyridine-3-carboxamide | |
| 91735 | |
| 394.30 | |
| Crystalline Powder |
| 161°C | |
| MFCD01311804 | |
| diflufenican, n-2,4-difluorophenyl-2-3-trifluoromethyl phenoxy nicotinamide, diflufenicanil, diflufenican bsi:iso, unii-3t8d1vdo6k, n-2,4-difluorophenyl-2-3-trifluoromethyl phenoxy pyridine-3-carboxamide, 3t8d1vdo6k, 2',4'-difluoro-2-alpha,alpha,alpha-trifluoro-m-tolyloxy nicotinanilide, n-2,4-difluorophenyl-2-3-trifluoromethyl phenoxy-3-pyridinecarboxamide | |
| C1=CC(=CC(=C1)OC2=C(C=CC=N2)C(=O)NC3=C(C=C(C=C3)F)F)C(F)(F)F | |
| 394.301 | |
| CHEBI:81824 | |
| ≥98.0% (HPLC,N) | |
| Diflufenican |
Safety and Handling
RTECSNumber : US4589800
TSCA : No