missing translation for 'onlineSavingsMsg'
Learn More

Difloxacin hydrochloride

Catalog No. AAJ6375406
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6375406 5 g
AAJ6375414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6375406 Supplier Thermo Scientific Chemicals Supplier No. J6375406

CAS: 91296-86-5 | C21H20ClF2N3O3 | 435.856 g/mol

A quinolone antibacterial compound. Structurally related to norfloxacin

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • A quinolone antibacterial compound
  • Structurally related to norfloxacin

Chemical Identifiers

CAS 91296-86-5
Molecular Formula C21H20ClF2N3O3
Molecular Weight (g/mol) 435.856
MDL Number MFCD03840489
InChI Key JFMGBGLSDVIOHL-UHFFFAOYSA-N
Synonym difloxacin hydrochloride, difloxacin hcl, unii-xj0260hj0o, abbott-56619, 6-fluoro-1-4-fluorophenyl-7-4-methylpiperazin-1-yl-4-oxo-1,4-dihydroquinoline-3-carboxylic acid hydrochloride, difloxacin hydrochloride usan, dsstox_cid_26599, dsstox_rid_81754, dsstox_gsid_46599
PubChem CID 56205
IUPAC Name 6-fluoro-1-(4-fluorophenyl)-7-(4-methylpiperazin-1-yl)-4-oxoquinoline-3-carboxylic acid;hydrochloride
SMILES CN1CCN(CC1)C2=C(C=C3C(=C2)N(C=C(C3=O)C(=O)O)C4=CC=C(C=C4)F)F.Cl

Specifications

Color White
Quantity 5 g
Beilstein 6464798
Merck Index 14,3141
Solubility Information Soluble in aqueous buffer
Formula Weight 435.85
Physical Form Powder
Chemical Name or Material Difloxacin hydrochloride
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only