missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Difco™ Yeast Extract, ultra-filtered (UF)
GREENER_CHOICE

Catalog No. DF210929
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DF210929 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. DF210929 Supplier Gibco™ Supplier No. 210929

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Difco™ Yeast Extracts are concentrates of the water-soluble portion of autolyzed Saccharomyces cerevisiae cells derived from primary grown baker's yeast.

  • Difco Yeast Extracts are concentrates of the water-soluble portion of autolyzed Saccharomyces cerevisiae cells derived from primary grown baker's yeast
  • These animal origin-free products are suitable for use as multi-functional nutritional supplements in mammalian cell culture, microbial fermentation, and insect cell culture applications
  • Yeast extract provides essential water-soluble vitamins, amino acids, peptides, and carbohydrates to any medium formulation
  • Difco Yeast Extract Low-Dusting (LD) was designed to limit the amount of dust generated compared to other spray-dried yeast extracts
  • It is ultra-filtered and specifically designed for mammalian cell culture applications
  • With its low endotoxin level and high content of naturally occurring B vitamins, it can be used effectively to reduce the need for fetal bovine serum
  • It has an endotoxin level of less than or equal to 500 EU/g
  • Store in cool, dry conditions in well-sealed containers at 2°C to 25°C

Specifications

Certifications/Compliance LUO
Content And Storage Storage conditions: 2°C to 25°C. Store in cool, dry conditions in well-sealed containers.
Cell Type Bacteria
Culture Type Mammalian, Insect, Microbial
Form Powder
Product Type Dehydrated Culture Media
Format Bottle
For Use With (Application) Cell culture and microbial fermentation, tissue culture application
Product Line Difco
Quantity 500 g
Source Animal origin-free
Show More Show Less