Learn More
BD Difco™ Poly Antisera
For agglutination test
Supplier: BD 228371
Description
Salmonella poly antiserum
- Febrile control antisera
- Helps in the detection of diseases such as salmonellosis, brucellosis and rickettsial diseases
- Qualitative and semi-quantitative slide or tube agglutination tests for homologous antibodies
- Used along with febrile antigens as positive or negative controls
- For identification of morphologically and biochemically identified Shigella species
- Can be used only in slide agglutination test
- Lyophilized, polyclonal rabbit antisera
Specifications
| For in vitro diagnostic use | |
| Shigella | |
| 3 mL |
For in vitro diagnostic use