Learn More
BD Difco™ Poly Antisera
For agglutination test
Supplier:  BD 227761
 View more versions of this product
Catalog No. DF2776-47-3
Only null left
Explore available promotions
Description
Description
Salmonella poly antiserum
  • Febrile control antisera
  • Helps in the detection of diseases such as salmonellosis, brucellosis and rickettsial diseases
  • Qualitative and semi-quantitative slide or tube agglutination tests for homologous antibodies
  • Used along with febrile antigens as positive or negative controls
Shigella antiserum poly group
  • For identification of morphologically and biochemically identified Shigella species
  • Can be used only in slide agglutination test
  • Lyophilized, polyclonal rabbit antisera
Specifications
Specifications
For in vitro diagnostic use
Shigella
3 mL
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Promotions
Promotions
For in vitro diagnostic use