missing translation for 'onlineSavingsMsg'
Learn More

Diethylzinc (ca. 17% in Hexane, ca. 1mol/L), TCI America™

Catalog No. D3214100ML
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D3214100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D3214100ML Supplier TCI America Supplier No. D3214100ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 557-20-0
Molecular Formula C4H10Zn
Molecular Weight (g/mol) 123.50
MDL Number MFCD00009021
InChI Key HQWPLXHWEZZGKY-UHFFFAOYSA-N
Synonym diethylzinc, zinc, diethyl, diethyl zinc, zinc ethide, zinc ethyl, unii-s0w5nqh7c6, et2zn, znet2, s0w5nqh7c6, c2h5 2zn
PubChem CID 11185
IUPAC Name diethylzinc
SMILES CC[Zn]CC

Specifications

Color Colorless
Physical Form Liquid
Quantity 100 mL
UN Number 3399
Formula Weight 123.50
Chemical Name or Material Diethylzinc (ca. 17% in Hexane, ca. 1mol/L)
EINECSNumber (9)-1890
RTECSNumber ZH2077777
TSCA Yes