missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diethylsilane, 96+%
Description
Diethylsilane is used as a precursor to prepare (3-allylsulfanyl-propyl)-diethyl-silane by using (Ph3P)3RhCl as reagent. It is also used in the chemical vapor deposition of SiO2 for microelectronics.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Diethylsilane is used as a precursor to prepare (3-allylsulfanyl-propyl)-diethyl-silane by using (Ph3P)3RhCl as reagent. It is also used in the chemical vapor deposition of SiO2 for microelectronics.
Solubility
Miscible with terahydrofuran, diethyl ether, 1,4-dioxane and benzene.
Notes
Moisture sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents, bases, oxygen and water.
Diethylsilane is used as a precursor to prepare (3-allylsulfanyl-propyl)-diethyl-silane by using (Ph3P)3RhCl as reagent. It is also used in the chemical vapor deposition of SiO2 for microelectronics.
Solubility
Miscible with terahydrofuran, diethyl ether, 1,4-dioxane and benzene.
Notes
Moisture sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents, bases, oxygen and water.
Specifications
Specifications
| Boiling Point | 56°C |
| Melting Point | -132°C |
| Quantity | 25 g |
| Flash Point | −20°C (−4°F) |
| Odor | Mild |
| Refractive Index | 1.392 |
| Sensitivity | Moisture sensitive |
| Density | 0.684 |
| UN Number | UN1993 |
| Beilstein | 1730902 |
| Show More |
Safety and Handling
| Hazard Category | H225-H315-H319-H335 |
| Hazard Statement | GHS H Statement H225-H315-H319-H335 Highly flammable liquid and vapor. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S. |
| EINECSNumber | 208-834-9 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures; Store under Nitrogen |
RUO – Research Use Only