accessibility menu, dialog, popup

UserName

CAS RN 542-91-6

IUPAC Name: diethylsilane
Synonyms: silane, diethyl-
Molecular Weight (g/mol): 88.23
Molecular Formula: C4H12Si
InChi Key: UCXUKTLCVSGCNR-UHFFFAOYSA-N
SMILES: CC[SiH2]CC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results