missing translation for 'onlineSavingsMsg'
Learn More

Diethylenetriaminepentaacetic Acid 98.0+%, TCI America™

Supplier:  TCI America D0504500G

 View more versions of this product

Catalog No. D0504500G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Diethylenetriaminepentaacetic Acid
220°C
C14H23N3O10
500 g
pentetic acid, diethylenetriaminepentaacetic acid, dtpa, detapac, complexon v, detarex, perma kleer, titriplex v, monaquest cai, hamp-ex acid
C(CN(CC(=O)O)CC(=O)O)N(CCN(CC(=O)O)CC(=O)O)CC(=O)O
393.349
CHEBI:35739
≥98.0% (T)
67-43-6
White
MFCD00004289
3077
QPCDCPDFJACHGM-UHFFFAOYSA-N
2-[bis[2-[bis(carboxymethyl)amino]ethyl]amino]acetic acid
3053
393.35
Crystalline Powder

Chemical Identifiers

67-43-6
393.349
QPCDCPDFJACHGM-UHFFFAOYSA-N
3053
2-[bis[2-[bis(carboxymethyl)amino]ethyl]amino]acetic acid
C14H23N3O10
MFCD00004289
pentetic acid, diethylenetriaminepentaacetic acid, dtpa, detapac, complexon v, detarex, perma kleer, titriplex v, monaquest cai, hamp-ex acid
CHEBI:35739
C(CN(CC(=O)O)CC(=O)O)N(CCN(CC(=O)O)CC(=O)O)CC(=O)O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-1273

RTECSNumber : MB8205000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions