missing translation for 'onlineSavingsMsg'
Learn More

Diethylenetriaminepentaacetic Acid 98.0+%, TCI America™

Catalog No. p-7126254
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D050425G 25 g
D0504500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D050425G Supplier TCI America Supplier No. D050425G
Only null left

Chemical Identifiers

CAS 67-43-6
Molecular Formula C14H23N3O10
Molecular Weight (g/mol) 393.349
MDL Number MFCD00004289
InChI Key QPCDCPDFJACHGM-UHFFFAOYSA-N
Synonym pentetic acid, diethylenetriaminepentaacetic acid, dtpa, detapac, complexon v, detarex, perma kleer, titriplex v, monaquest cai, hamp-ex acid
PubChem CID 3053
ChEBI CHEBI:35739
IUPAC Name 2-[bis[2-[bis(carboxymethyl)amino]ethyl]amino]acetic acid
SMILES C(CN(CC(=O)O)CC(=O)O)N(CCN(CC(=O)O)CC(=O)O)CC(=O)O

Specifications

Melting Point 220°C
Color White
pH 2.5 (10g/L H2O soln. 23°C)
Quantity 25 g
UN Number 3077
Formula Weight 393.35
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Diethylenetriaminepentaacetic Acid
EINECSNumber (2)-1273
RTECSNumber MB8205000
TSCA Yes