missing translation for 'onlineSavingsMsg'
Learn More

Diethylenetriaminepentaacetic Acid, Pentasodium Salt, 41% Solution in Water, Spectrum™ Chemical

Catalog No. 18602201
Change view
Click to view available options
Quantity:
100 mL
500 mL
4 L
Packaging:
Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18602201 100 mL Poly Bottle
18602202 500 mL Poly Bottle
18602203 4 L Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18602201 Supplier Spectrum Chemical Mfg Cor Supplier No. D1156100ML

D1156, 140-01-2, C14H18N3Na5O10

Spectrum™ Chemical Diethylenetriaminepentaacetic Acid, Pentasodium Salt, 41 Percent Solution in Water is also known under the abbreviation DETA and is a colorless liquid that is hygroscopic and soluble in both water and organic solvent that are polarized. Used as a tridentate ligand forming complex in coordination chemistry, it is also used for the extraction of gases that are acid in the oil industry.

Chemical Identifiers

CAS 140-01-2 , 7732-18-5
Molecular Formula C14H18N3Na5O10
Molecular Weight (g/mol) 503.26
MDL Number MFCD00051016
InChI Key LQPLDXQVILYOOL-UHFFFAOYSA-I
IUPAC Name pentasodium 2-[bis({2-[bis(carboxylatomethyl)amino]ethyl})amino]acetate
SMILES [Na+].[Na+].[Na+].[Na+].[Na+].[O-]C(=O)CN(CCN(CC([O-])=O)CC([O-])=O)CCN(CC([O-])=O)CC([O-])=O

Specifications

CAS Max % 41%, 59%
Packaging Poly Bottle
Quantity 100 mL
Percent Purity 39 to 41%
Grade Reagent