missing translation for 'onlineSavingsMsg'
Learn More

Diethylenetriamine 98.0+%, TCI America™

Supplier:  TCI America D049325ML

 View more versions of this product

Catalog No. D049325ML


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Diethylenetriamine
-35°C
209°C
MFCD00008171
2079
RPNUMPOLZDHAAY-UHFFFAOYSA-N
bis(2-aminoethyl)amine
8111
103.17
Liquid
111-40-0
Yellow
C4H13N3
25 mL
diethylenetriamine, bis 2-aminoethyl amine, 2,2'-diaminodiethylamine, diethylene triamine, barsamide 115, epicure t, ancamine deta, 1,4,7-triazaheptane, 2,2'-iminodiethylamine, n,n-bis 2-aminoethyl amine
NCCNCCN
103.17
CHEBI:30629
≥98.0% (GC,T)

Chemical Identifiers

111-40-0
103.17
RPNUMPOLZDHAAY-UHFFFAOYSA-N
8111
bis(2-aminoethyl)amine
C4H13N3
MFCD00008171
diethylenetriamine, bis 2-aminoethyl amine, 2,2'-diaminodiethylamine, diethylene triamine, barsamide 115, epicure t, ancamine deta, 1,4,7-triazaheptane, 2,2'-iminodiethylamine, n,n-bis 2-aminoethyl amine
CHEBI:30629
NCCNCCN
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0159

RTECSNumber : IE1225000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions