accessibility menu, dialog, popup

UserName

H2NNHNH2

CAS RN 111-40-0

IUPAC Name: bis(2-aminoethyl)amine
Synonyms: diethylenetriamine ChS-P 1 ethylamine, 2,2'-iminobis- barsamide 115 2,2'-iminobis-ethylamine diethylenetriamine diacetate 2,2'-iminodi(ethylamine) diethylenetriamine preparation dien diethylenetriamine (substance)
Molecular Weight (g/mol): 103.17
Molecular Formula: C4H13N3
InChi Key: RPNUMPOLZDHAAY-UHFFFAOYSA-N
SMILES: NCCNCCN
Boiling Point 
  • (3)
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (6)
  • (3)
  • (3)
  • (2)
  • (2)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (5)