missing translation for 'onlineSavingsMsg'
Learn More

Diethylenetriamine, 99%

Catalog No. AAA11033AE
Change view
Click to view available options
Quantity:
100 mL
500 mL
2500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA11033AE 100 mL
AAA11033AP 500 mL
AAA110330F 2500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA11033AE Supplier Thermo Scientific Chemicals Supplier No. A11033AE
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 111-40-0 | C4H13N3 | 103.17 g/mol

Diethylenetriamine is used as a solvent for plastics, dyes and in chemical synthesis. It serves as a curing agent for epoxy-resins. As a tridentate ligand, it form complexes such as Co(dien)(NO2)3 in coordination chemistry. It is also used as a fuel additive. It reacts with fatty acid resulting in the formation of amidoamines, which are used as corrosion inhibitors in the petroleum industry. It is associated with unsymmetrical dimethylhydrazine to prepare hydyne, which is a propellant for liquid-fuel rockets.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diethylenetriamine is used as a solvent for plastics, dyes and in chemical synthesis. It serves as a curing agent for epoxy-resins. As a tridentate ligand, it form complexes such as Co(dien)(NO2)3 in coordination chemistry. It is also used as a fuel additive. It reacts with fatty acid resulting in the formation of amidoamines, which are used as corrosion inhibitors in the petroleum industry. It is associated with unsymmetrical dimethylhydrazine to prepare hydyne, which is a propellant for liquid-fuel rockets.

Solubility
Miscible with water and ethanol. Immiscible with ether.

Notes
Air Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents, strong acids and copper.

Chemical Identifiers

CAS 111-40-0
Molecular Formula C4H13N3
Molecular Weight (g/mol) 103.17
MDL Number MFCD00008171
InChI Key RPNUMPOLZDHAAY-UHFFFAOYSA-N
Synonym diethylenetriamine, bis 2-aminoethyl amine, 2,2'-diaminodiethylamine, diethylene triamine, barsamide 115, epicure t, ancamine deta, 1,4,7-triazaheptane, 2,2'-iminodiethylamine, n,n-bis 2-aminoethyl amine
PubChem CID 8111
ChEBI CHEBI:30629
IUPAC Name N'-(2-aminoethyl)ethane-1,2-diamine
SMILES NCCNCCN

Specifications

Melting Point -40°C
Density 0.952
Boiling Point 200°C to 204°C
Flash Point 94°C (201°F)
Odor Ammonia-like
Refractive Index 1.483
Linear Formula H2NCH2CH2NHCH2CH2NH2
Quantity 100 mL
UN Number UN2079
Beilstein 605314
Sensitivity Air Sensitive
Solubility Information Miscible with water and ethanol. Immiscible with ether.
Formula Weight 103.17
Percent Purity 99%
Chemical Name or Material Diethylenetriamine
Show More Show Less
Hazard Category H227-H302+H312-H314-H317-H330-H335
Hazard Statement GHS H Statement
H314-H302-H312-H317
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful in contact with skin.
May cause an allergic skin reaction.
Precautionary Statement P210-P235-P260-P264b-P270-P271-P272-P280-P284-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: DIETHYLENETRIAMINE
EINECSNumber 203-865-4
RTECSNumber IE1225000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only