missing translation for 'onlineSavingsMsg'
Learn More

Diethylene Glycol Monophenyl Ether 98.0+%, TCI America™

$54.06 - $149.13

Chemical Identifiers

CAS 104-68-7
Molecular Formula C10H14O3
Molecular Weight (g/mol) 182.22
MDL Number MFCD00045989
InChI Key ZUAURMBNZUCEAF-UHFFFAOYSA-N
Synonym 2-(2-Phenoxyethoxy)ethanol, Phenyl Diglycol
PubChem CID 7715
IUPAC Name 2-(2-phenoxyethoxy)ethan-1-ol
SMILES OCCOCCOC1=CC=CC=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
D513925G
View Documents
TCI America
D513925G
25 g N/A
Only null left
 
D5139500G
View Documents
TCI America
D5139500G
500 g N/A
Only null left
 
Description

Description

Specifications

Chemical Identifiers

104-68-7
182.22
ZUAURMBNZUCEAF-UHFFFAOYSA-N
7715
OCCOCCOC1=CC=CC=C1
C10H14O3
MFCD00045989
2-(2-Phenoxyethoxy)ethanol, Phenyl Diglycol
2-(2-phenoxyethoxy)ethan-1-ol

Specifications

104-68-7
298°C
MFCD00045989
2-(2-Phenoxyethoxy)ethanol, Phenyl Diglycol
OCCOCCOC1=CC=CC=C1
182.22
182.22
Liquid
Yellow
C10H14O3
25 g
ZUAURMBNZUCEAF-UHFFFAOYSA-N
2-(2-phenoxyethoxy)ethan-1-ol
7715
≥98.0% (GC)
Diethylene Glycol Monophenyl Ether
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (3)-0563

RTECSNumber : KM0875000

TSCA : Yes

SDS
missing translation for 'documents'

missing translation for 'documents'