accessibility menu, dialog, popup

Test

CAS RN 104-68-7

CAS RN 104-68-7

IUPAC Name: 2-(2-phenoxyethoxy)ethan-1-ol
Synonyms: peg-2 phenyl ether
Molecular Weight (g/mol): 182.22
Molecular Formula: C10H14O3
InChi Key: ZUAURMBNZUCEAF-UHFFFAOYSA-N
SMILES: OCCOCCOC1=CC=CC=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results