missing translation for 'onlineSavingsMsg'
Learn More

Diethylene Glycol Monomethyl Ether (stabilized with BHT) 99.0+%, TCI America™

Catalog No. p-7132222
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M053725ML 25 mL
M0537500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M053725ML Supplier TCI America Supplier No. M053725ML
Only null left

Chemical Identifiers

CAS 111-77-3
Molecular Formula C5H12O3
Molecular Weight (g/mol) 120.15
MDL Number MFCD00002871,MFCD00084416
InChI Key SBASXUCJHJRPEV-UHFFFAOYSA-N
Synonym 2-2-methoxyethoxy ethanol, diethylene glycol monomethyl ether, methyl carbitol, methyl digol, methoxydiglycol, ethanol, 2-2-methoxyethoxy, methyl dioxitol, diethylene glycol methyl ether, dowanol dm, poly-solv dm
PubChem CID 8134
ChEBI CHEBI:44836
IUPAC Name 2-(2-methoxyethoxy)ethan-1-ol
SMILES COCCOCCO

Specifications

Melting Point -70°C
Color Colorless
Boiling Point 193°C
Quantity 25 mL
Formula Weight 120.15
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Diethylene Glycol Monomethyl Ether (stabilized with BHT)
EINECSNumber (2)-0422&(7)-0097
RTECSNumber KL6125000
TSCA Yes