missing translation for 'onlineSavingsMsg'
Learn More

Diethylene Glycol Monoethyl Ether Acetate 99.0+%, TCI America™

Supplier:  TCI America D0500500G

 View more versions of this product

Catalog No. D0500500G

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Diethylene Glycol Monoethyl Ether Acetate
Colorless
C8H16O4
500 g
FPZWZCWUIYYYBU-UHFFFAOYSA-N
2-(2-ethoxyethoxy)ethyl acetate
8165
≥99.0% (GC)
112-15-2
218°C
MFCD00041928
carbitol acetate, 2-2-ethoxyethoxy ethyl acetate, diethylene glycol monoethyl ether acetate, ektasolve de acetate, karbitolacetat, ethoxydiglycol acetate, glycol ether de acetate, diglycol monoethyl ether acetate, ethanol, 2-2-ethoxyethoxy-, acetate, ethyldiglycol acetate
CCOCCOCCOC(C)=O
176.21
176.21
Liquid

Chemical Identifiers

112-15-2
176.21
FPZWZCWUIYYYBU-UHFFFAOYSA-N
8165
CCOCCOCCOC(C)=O
C8H16O4
MFCD00041928
carbitol acetate, 2-2-ethoxyethoxy ethyl acetate, diethylene glycol monoethyl ether acetate, ektasolve de acetate, karbitolacetat, ethoxydiglycol acetate, glycol ether de acetate, diglycol monoethyl ether acetate, ethanol, 2-2-ethoxyethoxy-, acetate, ethyldiglycol acetate
2-(2-ethoxyethoxy)ethyl acetate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0744

RTECSNumber : KK8925000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions