Learn More
Diethylene glycol mono-n-hexyl ether, 97%
CAS: 112-59-4 | C10H22O3 | 190.28 g/mol
$87.38 - $942.67
Chemical Identifiers
| CAS | 112-59-4 |
|---|---|
| Molecular Formula | C10H22O3 |
| Molecular Weight (g/mol) | 190.28 |
| MDL Number | MFCD00010703 |
| InChI Key | GZMAAYIALGURDQ-UHFFFAOYSA-N |
| Synonym | diethylene glycol monohexyl ether, 2-2-hexyloxyethoxy ethanol, hexol carbitol, hexyl carbitol, n-hexyl carbitol, n-hexoxyethoxyethanol, 3,6-dioxadodecanol-1, ethanol, 2-2-hexyloxy ethoxy, hexyldiglycol, hexylkarbitol |
| PubChem CID | 8199 |
| IUPAC Name | 2-(2-hexoxyethoxy)ethanol |
| SMILES | CCCCCCOCCOCCO |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAB20774AP
|
Thermo Scientific Chemicals
B20774AP |
500 mL |
Each for $87.38
|
|
|||||
|
AAB207740F
|
Thermo Scientific Chemicals
B207740F |
2500 mL |
Each for $303.63
|
|
|||||
|
AAB207740D
|
Thermo Scientific Chemicals
B207740D |
10,000 mL |
Each for $942.67
|
|
|||||
Description
Used as a solvent for coatings and degreasing.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsUsed as a solvent for coatings and degreasing.
Solubility
Soluble in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Chemical Identifiers
| 112-59-4 | |
| 190.28 | |
| GZMAAYIALGURDQ-UHFFFAOYSA-N | |
| 8199 | |
| CCCCCCOCCOCCO |
| C10H22O3 | |
| MFCD00010703 | |
| diethylene glycol monohexyl ether, 2-2-hexyloxyethoxy ethanol, hexol carbitol, hexyl carbitol, n-hexyl carbitol, n-hexoxyethoxyethanol, 3,6-dioxadodecanol-1, ethanol, 2-2-hexyloxy ethoxy, hexyldiglycol, hexylkarbitol | |
| 2-(2-hexoxyethoxy)ethanol |
Specifications
| 112-59-4 | |
| 0.932 | |
| 123°C (253°F) | |
| 1.437 | |
| MFCD00010703 | |
| 1743959 | |
| Soluble in water. | |
| CCCCCCOCCOCCO | |
| 190.28 | |
| 190.28 | |
| Diethylene glycol mono-n-hexyl ether |
| -40°C | |
| 259°C to 261°C | |
| C10H22O3 | |
| HOCH2CH2OCH2CH2O(CH2)5CH3 | |
| 500 mL | |
| diethylene glycol monohexyl ether, 2-2-hexyloxyethoxy ethanol, hexol carbitol, hexyl carbitol, n-hexyl carbitol, n-hexoxyethoxyethanol, 3,6-dioxadodecanol-1, ethanol, 2-2-hexyloxy ethoxy, hexyldiglycol, hexylkarbitol | |
| GZMAAYIALGURDQ-UHFFFAOYSA-N | |
| 2-(2-hexoxyethoxy)ethanol | |
| 8199 | |
| 97% |
Safety and Handling
GHS H Statement
H318-H312
Causes serious eye damage.
Harmful in contact with skin.
P280-P302+P352-P305+P351+P338-P310-P312-P363-P501c
H312-H318
EINECSNumber : 203-988-3
RTECSNumber : KL2625000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only