missing translation for 'onlineSavingsMsg'
Learn More

Diethylene glycol mono-n-hexyl ether, 97%

CAS: 112-59-4 | C10H22O3 | 190.28 g/mol

Supplier:  Thermo Scientific Chemicals B207740F

 View more versions of this product

Catalog No. AAB207740F


Only null left
Explore available promotions

Description

Description

Used as a solvent for coatings and degreasing.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as a solvent for coatings and degreasing.

Solubility
Soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications

Specifications

Diethylene glycol mono-n-hexyl ether
-40°C
259°C to 261°C
C10H22O3
HOCH2CH2OCH2CH2O(CH2)5CH3
2500 mL
diethylene glycol monohexyl ether, 2-2-hexyloxyethoxy ethanol, hexol carbitol, hexyl carbitol, n-hexyl carbitol, n-hexoxyethoxyethanol, 3,6-dioxadodecanol-1, ethanol, 2-2-hexyloxy ethoxy, hexyldiglycol, hexylkarbitol
GZMAAYIALGURDQ-UHFFFAOYSA-N
2-(2-hexoxyethoxy)ethanol
8199
97%
112-59-4
0.932
123°C (253°F)
1.437
MFCD00010703
1743959
Soluble in water.
CCCCCCOCCOCCO
190.28
190.28

Chemical Identifiers

112-59-4
190.28
GZMAAYIALGURDQ-UHFFFAOYSA-N
8199
CCCCCCOCCOCCO
C10H22O3
MFCD00010703
diethylene glycol monohexyl ether, 2-2-hexyloxyethoxy ethanol, hexol carbitol, hexyl carbitol, n-hexyl carbitol, n-hexoxyethoxyethanol, 3,6-dioxadodecanol-1, ethanol, 2-2-hexyloxy ethoxy, hexyldiglycol, hexylkarbitol
2-(2-hexoxyethoxy)ethanol
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H318-H312
Causes serious eye damage.
Harmful in contact with skin.

P280-P302+P352-P305+P351+P338-P310-P312-P363-P501c

H312-H318

EINECSNumber : 203-988-3

RTECSNumber : KL2625000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only