missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diethylene Glycol Mono(2-propyn-1-yl) Ether 97.0+%, TCI America™
$397.05 - $397.05
Chemical Identifiers
| CAS | 7218-43-1 |
|---|---|
| Molecular Formula | C7H12O3 |
| Molecular Weight (g/mol) | 144.17 |
| MDL Number | MFCD00053353 |
| InChI Key | HUSDTFBXUYBZJD-UHFFFAOYSA-N |
| Synonym | 2-[2-(2-Propyn-1-yloxy)ethoxy]ethanol |
| PubChem CID | 81639 |
| IUPAC Name | 2-(2-prop-2-ynoxyethoxy)ethanol |
| SMILES | C#CCOCCOCCO |
Chemical Identifiers
| 7218-43-1 | |
| 144.17 | |
| HUSDTFBXUYBZJD-UHFFFAOYSA-N | |
| 81639 | |
| C#CCOCCOCCO |
| C7H12O3 | |
| MFCD00053353 | |
| 2-[2-(2-Propyn-1-yloxy)ethoxy]ethanol | |
| 2-(2-prop-2-ynoxyethoxy)ethanol |
Specifications
| 7218-43-1 | |
| 81°C | |
| MFCD00053353 | |
| 2-[2-(2-Propyn-1-yloxy)ethoxy]ethanol | |
| C#CCOCCOCCO | |
| 144.17 | |
| 144.17 | |
| Liquid |
| Yellow | |
| C7H12O3 | |
| 1 g | |
| HUSDTFBXUYBZJD-UHFFFAOYSA-N | |
| 2-(2-prop-2-ynoxyethoxy)ethanol | |
| 81639 | |
| ≥97.0% (GC) | |
| Diethylene Glycol Mono(2-propyn-1-yl) Ether |
Safety and Handling
EINECSNumber : (2)-2398
TSCA : No