accessibility menu, dialog, popup

UserName

CAS RN 7218-43-1

HOOOCH

CAS RN 7218-43-1

IUPAC Name: 2-[2-(prop-2-yn-1-yloxy)ethoxy]ethan-1-ol
Synonyms: 2-[2-(prop-2-yn-1-yloxy)ethoxy]ethanol
Molecular Weight (g/mol): 144.17
Molecular Formula: C7H12O3
InChi Key: HUSDTFBXUYBZJD-UHFFFAOYSA-N
SMILES: OCCOCCOCC#C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results