missing translation for 'onlineSavingsMsg'
Learn More

Diethylene glycol divinyl ether, 98%, stab. 0.1% potassium hydroxide

Catalog No. AAL1250118
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1250118 50 g
AAL1250109 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1250118 Supplier Thermo Scientific Chemicals Supplier No. L1250118
Only null left

CAS: 764-99-8 | C8H14O3 | 158.197 g/mol

Diethylene glycol divinyl ether is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diethylene glycol divinyl ether is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Insoluble in water.

Notes
Light Sensitive. Store in the dark. Incompatible with oxidizing agents and light.

Chemical Identifiers

CAS 764-99-8
Molecular Formula C8H14O3
Molecular Weight (g/mol) 158.197
MDL Number MFCD00054610
InChI Key SAMJGBVVQUEMGC-UHFFFAOYSA-N
Synonym diethylene glycol divinyl ether, divinylcarbitol, dvedeg, dvedeg russian, ether, bis 2-vinyloxy ethyl, ethene, 1,1'-oxybis 2,1-ethanediyloxy bis, 3,6,9-trioxaundeca-1,10-diene, unii-abm90y2g95, divinyl ether diethylenglykolu, bis 2-vinyloxy ethyl ether
PubChem CID 12998
IUPAC Name 1-ethenoxy-2-(2-ethenoxyethoxy)ethane
SMILES C=COCCOCCOC=C

Specifications

Melting Point -21°C
Density 0.968
Boiling Point 198°C to 199°C
Flash Point 83°C (181°F)
Refractive Index 1.447
Quantity 50 g
Beilstein 1752316
Sensitivity Light sensitive
Solubility Information Insoluble in water.
Formula Weight 158.2
Percent Purity 98%
Chemical Name or Material Diethylene glycol divinyl ether, stabilized 0.1% potassium hydroxide
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H227
Causes skin irritation.
Causes serious eye irritation.
Combustible liquid.
Precautionary Statement GHS P Statement
P210-P280-P305+P351+P338-P362-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Take off contaminated clothing and wash before reuse.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 212-133-3
RTECSNumber KN3850000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only