missing translation for 'onlineSavingsMsg'
Learn More

Diethylene glycol dimethyl ether, 99%, stab. with 100ppm BHT

CAS: 111-96-6 | C6H14O3 | 134.18 g/mol

Supplier:  Thermo Scientific Chemicals A133970F

 View more versions of this product

Catalog No. AAA133970F


Only null left
Explore available promotions

Description

Description

Diethylene glycol dimethyl ether is used as a solvent in organic reactions due to its stability towards higher pH and its high boiling point. It is particularly involved in reactions utilizing organometallic reagents such as Grignard reactions and metal hydride reductions. It is also a solvent for hydroboration reactions with diborane.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diethylene glycol dimethyl ether is used as a solvent in organic reactions due to its stability towards higher pH and its high boiling point. It is particularly involved in reactions utilizing organometallic reagents such as Grignard reactions and metal hydride reductions. It is also a solvent for hydroboration reactions with diborane.

Solubility
Miscible with water, ethanol, acetone, acetic acid, glycerine, pyridine and aldehydes. Slightly miscible with ether.

Notes
Hygroscopic. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
Specifications

Specifications

Diethylene glycol dimethyl ether
111-96-6
0.944
51°C (124°F)
C6H14O3
(CH3OCH2CH2)2O
2500 mL
1736101
14,3165
Miscible with water,ethanol,acetone,acetic acid,glycerine,pyridine and aldehydes. Slightly miscible with ether.
COCCOCCOC
134.175
CHEBI:46784
99%
stabilized with 100ppm BHT
-64°C
161°C to 162°C
Ether-like
1.408
MFCD00008503
UN3271
Hygroscopic
diglyme, bis 2-methoxyethyl ether, 2-methoxyethyl ether, 1-methoxy-2-2-methoxyethoxy ethane, diethylene glycol dimethyl ether, poly-solv, di 2-methoxyethyl ether, dimethyl carbitol, 2,5,8-trioxanonane
SBZXBUIDTXKZTM-UHFFFAOYSA-N
1-methoxy-2-(2-methoxyethoxy)ethane
8150
134.18

Chemical Identifiers

111-96-6
MFCD00008503
diglyme, bis 2-methoxyethyl ether, 2-methoxyethyl ether, 1-methoxy-2-2-methoxyethoxy ethane, diethylene glycol dimethyl ether, poly-solv, di 2-methoxyethyl ether, dimethyl carbitol, 2,5,8-trioxanonane
CHEBI:46784
COCCOCCOC
C6H14O3
SBZXBUIDTXKZTM-UHFFFAOYSA-N
8150
1-methoxy-2-(2-methoxyethoxy)ethane
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H360-H226
May damage fertility or the unborn child.
Flammable liquid and vapour.

P201-P202-P210-P233-P235-P240-P241-P242-P243-P260-P271-P281-P303+P361+P353-P304+P340-P308+P313-P370+P378q-P501c

H226-H336-H360FD-H373

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ETHERS, N.O.S.

EINECSNumber : 203-924-4

RTECSNumber : KN3339000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only