missing translation for 'onlineSavingsMsg'
Learn More

Diethylene Glycol Diacetate 98.0+%, TCI America™

Catalog No. D123025ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D123025ML 25 mL
D1230500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D123025ML Supplier TCI America Supplier No. D123025ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 628-68-2
Molecular Formula C8H14O5
Molecular Weight (g/mol) 190.195
MDL Number MFCD00026211
InChI Key UBPGILLNMDGSDS-UHFFFAOYSA-N
Synonym Oxybis(2-ethyl Acetate)
PubChem CID 69411
ChEBI CHEBI:87344
IUPAC Name 2-(2-acetyloxyethoxy)ethyl acetate
SMILES CC(=O)OCCOCCOC(=O)C

Specifications

Color Yellow
Boiling Point 250°C
Quantity 25 mL
Formula Weight 190.20
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Diethylene Glycol Diacetate
EINECSNumber (2)-0751
RTECSNumber AJ1800000
TSCA Yes