accessibility menu, dialog, popup

UserName

CAS RN 628-68-2

IUPAC Name: 2-[2-(acetyloxy)ethoxy]ethyl acetate
Synonyms: 2-[2-(acetyloxy)ethoxy]ethyl acetate
Molecular Weight (g/mol): 190.2
Molecular Formula: C8H14O5
InChi Key: UBPGILLNMDGSDS-UHFFFAOYSA-N
SMILES: CC(=O)OCCOCCOC(C)=O
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results