missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diethylene Glycol Bis(p-toluenesulfonate) 96.0+%, TCI America™
$78.29 - $253.51
Chemical Identifiers
| CAS | 7460-82-4 |
|---|---|
| Molecular Formula | C18H22O7S2 |
| Molecular Weight (g/mol) | 414.49 |
| MDL Number | MFCD00026001 |
| InChI Key | VYVPNTJBGPQTFA-UHFFFAOYSA-N |
| Synonym | Bis[2-(p-toluenesulfonyloxy)ethyl] Ether, Bis(2-tosyloxyethyl) Ether, Diethylene Glycol Ditosylate |
| PubChem CID | 316659 |
| IUPAC Name | 2-{2-[(4-methylbenzenesulfonyl)oxy]ethoxy}ethyl 4-methylbenzene-1-sulfonate |
| SMILES | CC1=CC=C(C=C1)S(=O)(=O)OCCOCCOS(=O)(=O)C1=CC=C(C)C=C1 |
Chemical Identifiers
| 7460-82-4 | |
| 414.49 | |
| VYVPNTJBGPQTFA-UHFFFAOYSA-N | |
| 316659 | |
| CC1=CC=C(C=C1)S(=O)(=O)OCCOCCOS(=O)(=O)C1=CC=C(C)C=C1 |
| C18H22O7S2 | |
| MFCD00026001 | |
| Bis[2-(p-toluenesulfonyloxy)ethyl] Ether, Bis(2-tosyloxyethyl) Ether, Diethylene Glycol Ditosylate | |
| 2-{2-[(4-methylbenzenesulfonyl)oxy]ethoxy}ethyl 4-methylbenzene-1-sulfonate |
Specifications
| 7460-82-4 | |
| C18H22O7S2 | |
| 5 g | |
| VYVPNTJBGPQTFA-UHFFFAOYSA-N | |
| 2-{2-[(4-methylbenzenesulfonyl)oxy]ethoxy}ethyl 4-methylbenzene-1-sulfonate | |
| 316659 | |
| ≥96.0% (HPLC,T) | |
| Diethylene Glycol Bis(p-toluenesulfonate) |
| 87°C | |
| MFCD00026001 | |
| Bis[2-(p-toluenesulfonyloxy)ethyl] Ether, Bis(2-tosyloxyethyl) Ether, Diethylene Glycol Ditosylate | |
| CC1=CC=C(C=C1)S(=O)(=O)OCCOCCOS(=O)(=O)C1=CC=C(C)C=C1 | |
| 414.49 | |
| 414.49 | |
| Crystalline Powder |
Safety and Handling
TSCA : No