accessibility menu, dialog, popup

UserName

CH3SOOOOOSOOH3C

CAS RN 7460-82-4

IUPAC Name: 2-{2-[(4-methylbenzenesulfonyl)oxy]ethoxy}ethyl 4-methylbenzene-1-sulfonate
Synonyms: 2-{2-[(4-methylbenzenesulfonyl)oxy]ethoxy}ethyl 4-methylbenzenesulfonate
Molecular Weight (g/mol): 414.49
Molecular Formula: C18H22O7S2
InChi Key: VYVPNTJBGPQTFA-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)S(=O)(=O)OCCOCCOS(=O)(=O)C1=CC=C(C)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results